(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

26411-0010 26411-2500
TS26411-0010EA 423 USD
TS26411-0010 TS26411-2500
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formula: C₁₀H₈ClF₃O₂
Boiling Pt: 213…214 °C (760 mmHg)
Density: 1.34 g/cm³
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067105
CAS Number: 20445-33-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4700 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +130° to +134° (20°C, 589 nm) (c=6, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR