Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

43865-0010 43865-0050
TS43865-0010EA 323 USD
TS43865-0010 TS43865-0050
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) 95%
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)
Formula: C₄H₄NNaO₆S
Melting Pt: 250 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Sodium (Na) 9.95 to 11.22 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR