2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP

36864-0250 36864-0010 36864-0050 36864-1000
TS36864-0250EA 296 USD
TS36864-0250 TS36864-0010 TS36864-0050 TS36864-1000
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
Melting Pt: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 98327-87-8

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 280 to 285 °C
NMR (31P-NMR) ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR