Cresyl violet (acetate), pure, certified

Supplier: Thermo Scientific Chemicals
Warning

40576-0100 40576-0500 40576-0025
TS40576-0100EA 186 USD
TS40576-0100 TS40576-0500 TS40576-0025
Cresyl violet (acetate), pure, certified
Cresyl violet (acetate)
Formula: C₁₈H₁₅N₃O₃
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00013151
CAS Number: 10510-54-0
EINECS: 208-953-6
Merck Index: 13,04406

Order Now

Specification Test Results

Appearance Dark green to black Crystalline powder
Infrared spectrum Conforms
Dye content ≥65 %
UV Lambda max 596 to 599 nm
Absorption Ratio P-15/P+15 0.94 to 1.2
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR