Formic acid-[D2] 95% (99% D) in D₂O for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

23340-0010 23340-0100
TS23340-0010EA 78 USD
TS23340-0010 TS23340-0100
Formic acid-[D2] 95% (99% D) in D₂O for NMR spectroscopy
Formic acid-[D2]
Formula: DCO₂D
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 5.9 °C
Density: 1.27 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037363
CAS Number: 920-42-3
UN: 1779
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.0 %
Titration with NaOH ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR