m-Xylylenediamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Benzenedimethanamine

14102-5000 14102-0050 14102-1000
TS14102-5000EA 139 USD
TS14102-5000 TS14102-0050 TS14102-1000
m-Xylylenediamine 99%
m-Xylylenediamine
Formula: C₈H₁₂N₂
MW: 136.1943 g/mol
Boiling Pt: 265 °C (745 mmHg)
Density: 1.03 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008119
CAS Number: 1477-55-0
EINECS: 216-032-5
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5720 (20°C, 589 nm)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR