Ferrozine mono-sodium salt hydrate 98+%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: FerroZine iron reagent hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate, 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate, PDT disulphonate monosodium salt hydrate

41057-0050 41057-0250 41057-0010
TS41057-0050EA 217 USD
TS41057-0050 TS41057-0250 TS41057-0010
Ferrozine mono-sodium salt hydrate 98+%, pure
Ferrozine mono-sodium salt hydrate
Formula: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150794
CAS Number: 63451-29-6

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Specific extinction E (1%/1cm) ≤0.015 (562 nm in water)
Titration with NaOH ≥98.0 % (On dry substance)
Suitability (for Cu + Fe determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR