Dodecacarbonyltriruthenium 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Triruthenium dodecacarbonyl, Ruthenium carbonyl

22400-0010 AC22400-0050
TS22400-0010EA 140 USD
TS22400-0010 TS22400-0050
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium
Formula: C₁₂O₁₂Ru₃
MW: 639.33 g/mol
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00011209
CAS Number: 15243-33-1
EINECS: 239-287-4

Order Now

Specification Test Results

Appearance Orange Crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR