Tris(pentafluorophenyl)borane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tris(perfluorophenyl)borane
35313-0050
35313-0010
TS35313-0050EA
564
USD
TS35313-0050
TS35313-0010
Tris(pentafluorophenyl)borane 95%
Tris(pentafluorophenyl)borane
Formula:
C₁₈BF₁₅ MW: 511.98 g/mol Melting Pt: 126…131 °C Storage Temperature: Freezer |
MDL Number:
MFCD00269813 CAS Number: 1109-15-5 EINECS: 213-504-2 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...