2,3-Diphenylmaleic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Diphenylfuran-2,5-dione

43901-0250 43901-0010 43901-0050
TS43901-0250EA 724 USD
TS43901-0250 TS43901-0010 TS43901-0050
2,3-Diphenylmaleic anhydride 98%
2,3-Diphenylmaleic anhydride
Formula: C₁₆H₁₀O₃
Melting Pt: 157.1…159.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005521
CAS Number: 4808-48-4

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 156 to 162 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR