Bis(pinacolato)diboron 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

33057-0250 33057-0010 33057-0050 33057-1000
TS33057-0250EA 214 USD
TS33057-0250 TS33057-0010 TS33057-0050 TS33057-1000
Bis(pinacolato)diboron 98%
Bis(pinacolato)diboron
Formula: C₁₂H₂₄B₂O₄
Melting Pt: 135…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now

Specification Test Results

Appearance White Powder or platelets
Infrared spectrum Conforms
Melting point 135 to 140 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR