Copper(I) bromide dimethyl sulfide complex 99%
Supplier: Thermo Scientific Chemicals
21318-0250
AC21318-0100
AC21318-1000
TS21318-0250EA
162
USD
TS21318-0250
TS21318-0100
TS21318-1000
Copper(I) bromide dimethyl sulfide complex 99%
Copper (I) bromide dimethyl sulphide complex
Formula:
C₂H₆BrCuS Melting Pt: 132 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043295 CAS Number: 54678-23-8 EINECS: 259-292-5 |
Specification Test Results
Appearance | Off-white to beige to green to gray Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Titration Iodometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...