Zinc (II) phthalocyanine (Sulfate ionophore II) 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex
42275-0250
AC42275-0050
TS42275-0250EA
181
USD
TS42275-0250
TS42275-0050
Zinc (II) phthalocyanine (Sulfate ionophore II) 96%
Zinc (II) phthalocyanine (Sulphate ionophore II)
Formula:
C₃₂H₁₆N₈Zn MW: 577.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041955 CAS Number: 14320-04-8 EINECS: 238-262-5 |
Specification Test Results
Appearance | Blue to purple Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % (105°C) |
UV spectrophotometry | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...