Methyl vinyl sulfone 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

30673-0500 30673-0010 30673-0100
TS30673-0500EA 679 USD
TS30673-0500 TS30673-0010 TS30673-0100
Methyl vinyl sulfone 95% stabilized
Methyl vinyl sulphone
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 115…117 °C (19 mmHg)
Density: 1.21 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007567
CAS Number: 3680-02-2
EINECS: 222-958-0
UN: 2810
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4650 (20°C, 589 nm)
GC ≥94 %
Stabilizer 200 ppm 4-tert-Butylphenol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR