Cyclopentadienyltitanium trichloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
21577-0100
AC21577-0025
TS21577-0100EA
240
USD
TS21577-0100
TS21577-0025
Cyclopentadienyltitanium trichloride 99%
Cyclopentadienyltitanium trichloride
Formula:
C₅Cl₃Ti Melting Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003724 CAS Number: 1270-98-0 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Yellow to orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...