Digitonin

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Digitonin

40756-0050 40756-5000
TS40756-0050EA 1719 USD
TS40756-0050 TS40756-5000
Digitonin
Digitonin
Formula: C₅₆H₉₂O₂₉
MW: 1229.312 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00077729
CAS Number: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Loss on drying ≤6 % (1 g, 105°C)
Appearance of solution (2.5% in warm EtOH) clear colorless to faint yellow
Residue after ignition ≤0.3 %
Specific optical rotation -45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR