3-Hydroxyphthalic anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxyisobenzofuran-1,3-dione

39435-0050 39435-0010
TS39435-0050EA 776 USD
TS39435-0050 TS39435-0010
3-Hydroxyphthalic anhydride 97%
3-Hydroxyphthalic anhydride
Formula: C₈H₄O₄
MW: 164.1150 g/mol
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00011557
CAS Number: 37418-88-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Powder or crystals or flakes
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR