tert-Butyldimethylsilyl trifluoromethanesulphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: t-Butyldimethylsilyl trifluoromethanesulphonate

20986-0050 20986-0250
TS20986-0050EA 91 USD
TS20986-0050 TS20986-0250
tert-Butyldimethylsilyl trifluoromethanesulphonate 98%
tert-Butyldimethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 65…67 °C (12 mmHg)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000405
CAS Number: 69739-34-0
EINECS: 274-102-0
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3880 (20°C, 589 nm)
Assay ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR