3-Nitrostyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

12899-0100 12899-0025
TS12899-0100EA 291 USD
TS12899-0100 TS12899-0025
3-Nitrostyrene 97% stabilized
3-Nitrostyrene
Formula: C₈H₇NO₂
Melting Pt: –5 °C
Density: 1.07 g/cm³
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007276
CAS Number: 586-39-0
EINECS: 209-575-4

Order Now

Specification Test Results

Appearance Clear yellow to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5840 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Approx. 0.1 % 3,5-di-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR