O-Methylhydroxylamine hydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
21049-0250
21049-1000
AC21049-0050
TS21049-0250EA
256
USD
TS21049-0250
TS21049-1000
TS21049-0050
O-Methylhydroxylamine hydrochloride 98+%
O-Methylhydroxylamine hydrochloride
Formula:
CH₆ClNO Melting Pt: 148…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012951 CAS Number: 593-56-6 EINECS: 209-798-7 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 148 to 153 °C |
Titration with HClO4 | 98.0 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...