Alcian blue 8GX, certified
Supplier: Thermo Scientific Chemicals
40046-0100
40046-0250
40046-1000
TS40046-0100EA
184
USD
TS40046-0100
TS40046-0250
TS40046-1000
Alcian blue 8GX, certified
Alcian blue 8 GX
Formula:
C₅₆H₆₈Cl₄CuN₁₆S₄ Melting Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010720 CAS Number: 33864-99-2 EINECS: 251-705-7 |
Specification Test Results
Appearance | Dark blue to purple Powder |
Infrared spectrum | Conforms |
Dye content | ≥50 % |
UV Lambda max | 615 to 635 nm (in water) |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...