1,2-Bis(dichlorophosphino)ethane 97%

Supplier: Thermo Scientific Chemicals
Danger

32147-0050 32147-0010
TS32147-0050EA 254 USD
TS32147-0050 TS32147-0010
1,2-Bis(dichlorophosphino)ethane 97%
1,2-Bis(dichlorophosphino)ethane
Formula: C₂H₄Cl₄P₂
MW: 231.813 g/mol
Boiling Pt: 70 °C (1 mmHg)
Density: 1.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009609
CAS Number: 28240-69-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5875 to 1.5935 (20°C, 589 nm)
NMR ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR