Dipropyleneglycol monomethyl ether (mixture of isomers) 99%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Dipropylene glycol monomethyl ether, 1-((1-Methoxypropan-2-yl)oxy)propan-2-ol
42820-0050
428200010
TS42820-0050EA
166
USD
TS42820-0050
TS428200010
Dipropyleneglycol monomethyl ether (mixture of isomers) 99%, pure
Dipropyleneglycol monomethyl ether (mixture of isomers)
Formula:
C₇H₁₆O₃ MW: 148.2001 g/mol Boiling Pt: 180 °C (760 mmHg) Melting Pt: –80 °C Density: 0.95 g/cm³ Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059604 CAS Number: 34590-94-8 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥98.5 % sum of isomers |
Elemental Analysis | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...