Isoquinoline-8-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isoquinolin-8-amine, 8-Aminoisoquinoline

42940-0050 42940-0010
TS42940-0050EA 442 USD
TS42940-0050 TS42940-0010
Isoquinoline-8-amine 97%
Isoquinoline-8-amine
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00179553
CAS Number: 23687-27-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 167 to 170 °C
GC ≥96.0 %
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR