2-Methyl-2-oxazoline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methyl-4,5-dihydrooxazole

15997-5000 15997-1000
TS15997-5000EA 322 USD
TS15997-5000 TS15997-1000
2-Methyl-2-oxazoline 99%
2-Methyl-2-oxazoline
Formula: C₄H₇NO
MW: 85.11 g/mol
Density: 1 g/cm³
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00005298
CAS Number: 1120-64-5
EINECS: 214-316-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC 1.5 to 2.5 % (MEK)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR