Bis(2-ethylhexyl)phosphate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

40247-0050 40247-0010
TS40247-0050EA 504 USD
TS40247-0050 TS40247-0010
Bis(2-ethylhexyl)phosphate 95%
Bis(2-ethylhexyl)phosphate
Formula: C₁₆H₃₅O₄P
MW: 322.43 g/mol
Boiling Pt: 41 °C (760 mmHg)
Melting Pt: –115 °C
Density: 0.65 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009492
CAS Number: 298-07-7
EINECS: 209-195-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3750 to 1.3780 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR