1-Aminobenzotriazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Benzo[d][1,2,3]triazol-1-amine

34864-2500 34864-1000
TS34864-2500EA 226 USD
TS34864-2500 TS34864-1000
1-Aminobenzotriazole 98%
1-Aminobenzotriazole
Formula: C₆H₆N₄
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00132902
CAS Number: 1614-12-6

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR