(±)-2,3-Epoxy-1-propanol 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Glycidol
12005-5000
12005-0025
12005-1000
TS12005-5000EA
211
USD
TS12005-5000
TS12005-0025
TS12005-1000
(±)-2,3-Epoxy-1-propanol 96%
(±)-2,3-Epoxy-1-propanol
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 160…161 °C (760 mmHg) Melting Pt: –54 °C Density: 1.11 g/cm³ Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005147 CAS Number: 556-52-5 EINECS: 209-128-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4360 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...