(±)-Allyl-2,3-epoxypropyl ether 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Allyl glycidyl ether

10294-5000 10294-0025 10294-1000
TS10294-5000EA 96 USD
TS10294-5000 TS10294-0025 TS10294-1000
(±)-Allyl-2,3-epoxypropyl ether 99+%
(±)-Allyl-2,3-epoxypropyl ether
Formula: C₆H₁₀O₂
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –100 °C
Density: 0.962 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005143
CAS Number: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4332 to 1.4352 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR