(Triphenylphosphine)gold(I) chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

39161-0050 AC39161-0010 AC39161-2500
TS39161-0050EA 699 USD
TS39161-0050 TS39161-0010 TS39161-2500
(Triphenylphosphine)gold(I) chloride 99%
(Triphenylphosphine)gold(I) chloride
Formula: C₁₈H₁₅AuClP
MW: 494.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009588
CAS Number: 14243-64-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥98.5 % (ex Au)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR