4,5-Dichlorophthalic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 4,5-Dichlorophthalic Anhydride
38712-0250
38712-0050
TS38712-0250EA
267
USD
TS38712-0250
TS38712-0050
4,5-Dichlorophthalic anhydride 98%
4,5-Dichlorophthalic anhydride
Formula:
C₈H₂Cl₂O₃ MW: 217.01 g/mol Melting Pt: 185…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075034 CAS Number: 942-06-3 |
Specification Test Results
Appearance | White to yellow to grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 185 to 189 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...