Bis(2-ethylhexyl)adipate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate

40246-0025 40246-0010 40246-2500
TS40246-0025EA 108 USD
TS40246-0025 TS40246-0010 TS40246-2500
Bis(2-ethylhexyl)adipate 99%
Bis(2-ethylhexyl)adipate
Formula: C₂₂H₄₂O₄
Boiling Pt: 210…218 °C (5 mmHg)
Melting Pt: –76 °C
Density: 0.924 g/cm³
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4500 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR