p-Anisidinium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Anisidine hydrochloride, 4-Methoxyaniline hydrochloride

40130-0250 40130-1000 40130-0050
TS40130-0250EA 124 USD
TS40130-0250 TS40130-1000 TS40130-0050
p-Anisidinium chloride 99%
p-Anisidinium chloride
Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00036388
CAS Number: 20265-97-8
EINECS: 243-657-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to pink to gray-brown Crystalline powder
Infrared spectrum Conforms
Melting point 214 to 219 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR