Sulfamide 99%

Supplier: Thermo Scientific Chemicals
Danger

19902-1000 19902-0050 19902-0250
TS19902-1000EA 579 USD
TS19902-1000 TS19902-0050 TS19902-0250
Sulfamide 99%
Sulphamide
Formula: H₄N₂O₂S
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.92 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00011606
CAS Number: 7803-58-9
EINECS: 203-562-7
UN: 2403
ADR: 3,II
Merck Index: 13,05223

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥98.5 %
Elemental Analysis ≥98.0 %
Related substances (TLC) Passes test
pH 5.0 to 7.0 (10 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR