2-Hydroxyethyl macrylate 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxyethyl acrylate, Acrylic acid 2-hydroxyethyl ester

33408-2500 33408-0010 33408-0025
TS33408-2500EA 64 USD
TS33408-2500 TS33408-0010 TS33408-0025
2-Hydroxyethyl macrylate 99+% stabilized
2-Hydroxyethyl macrylate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 78…80 °C (760 mmHg)
Melting Pt: –109.9 °C
Density: 0.7 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00002865
CAS Number: 818-61-1
EINECS: 209-845-1
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4015 to 1.4035 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤30 APHA
Stabilizer 200 to 450 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR