Butyl acrylate 92% tech. stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Butyl acrylate, Acrylic acid butyl ester

21586-0010 21586-0025 21586-0100
TS21586-0010EA 73 USD
TS21586-0010 TS21586-0025 TS21586-0100
Butyl acrylate 92% tech. stabilized
Butyl acrylate
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 3…5 °C
Density: 0.783 g/cm³
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 211-105-8
UN: 2348
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
GC ≥90 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Acidity ≤0.01 % (as Acrylic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR