2-Ethylhexyl acrylate 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acrylic acid 2-ethylhexyl ester

41011-0025 41011-0010 41011-0250
TS41011-0025EA 125 USD
TS41011-0025 TS41011-0010 TS41011-0250
2-Ethylhexyl acrylate 97% stabilized
2-Ethylhexyl acrylate
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 54 °C (1013 hPa)
Melting Pt: –155 °C
Density: 0.66 g/cm³
Flash Pt: –31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009495
CAS Number: 103-11-7
EINECS: 211-720-1
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3810 to 1.3830 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Acidity ≤0.01 % (as acrylic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR