trans,trans-2,4-Hexadien-1-ol 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(2E,4E)-Hexa-2,4-dien-1-ol, Trans,trans-2,4-hexadien-1-ol, (2E,4E)-1-Hydroxyhexa-2,4-diene, Sorbic alcohol, trans,trans-Hexa-2,4-dien-1-ol
32557-0500
32557-0100
32557-2500
TS32557-0500EA
237
USD
TS32557-0500
TS32557-0100
TS32557-2500
trans,trans-2,4-Hexadien-1-ol 99% stabilised
trans,trans-2,4-Hexadien-1-ol
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 80 °C (12 mmHg) Melting Pt: 30.5…31.5 °C Density: 0.87 g/cm³ Flash Pt: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002925 CAS Number: 17102-64-6 EINECS: 241-173-4 UN: 2811 ADR: 6.1 III |
Specification Test Results
Appearance | Clear colorless to white to light yellow Low melting solid or liquid after melting |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
Stabilizer | 0.1 % alpha-tocopherol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...