3,4-Dibutoxy-3-cyclobutene-1,2-dione 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Squaric acid dibutylester, Dibutyl squarate, 3,4-Dibutoxycyclobut-3-ene-1,2-dione, 1,2-Dibutoxy-1-cyclobutene-3,4-dione

13317-0050 13317-0250 13317-0010
TS13317-0050EA 94 USD
TS13317-0050 TS13317-0250 TS13317-0010
3,4-Dibutoxy-3-cyclobutene-1,2-dione 97%
3,4-Dibutoxy-3-cyclobutene-1,2-dione
Formula: C₁₂H₁₈O₄
MW: 226.27 g/mol
Boiling Pt: 121
Density: 1.05 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00037150
CAS Number: 2892-62-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥96.0 %
NMR (13C-NMR) Consistent with structure
NMR (31P-NMR) Consistent with structure
Elemental Analysis 76.56 to 82.94 % C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR