(S)-(+)-Carvone (synthetic) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

37784-1000 37784-0250
TS37784-1000EA 154 USD
TS37784-1000 TS37784-0250
(S)-(+)-Carvone (synthetic) 98%
(S)-(+)-Carvone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 230 °C (760 mmHg)
Density: 0.956 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00062997
CAS Number: 2244-16-8
EINECS: 218-827-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.5010 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.955 to 0.960
Specific optical rotation +52.0° to +62.0° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR