Osmium(III) chloride hydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Osmium trichloride hydrate

31870-0025 AC31870-5000
TS31870-0025EA 613 USD
TS31870-0025 TS31870-5000
Osmium(III) chloride hydrate
Osmium(III) chloride hydrate
Formula: Cl₃Os·H₂O
MW: 314.6
Melting Pt: >500 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149815
CAS Number: 14996-60-2
EINECS: 236-587-7
UN: 2923
ADR: 8 III

Order Now

Specification Test Results

Appearance Dark red to black Powder or crystals
Osmium (Os) 53 to 56 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR