Methylmagnesium bromide 3 M in diethyl ether, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
18354-1000
18354-8000
TS18354-1000EA
79
USD
TS18354-1000
TS18354-8000
Methylmagnesium bromide 3 M in diethyl ether, AcroSeal™
Methylmagnesium bromide
Formula:
CH₃BrMg MW: 119.24 g/mol Density: 1.035 g/cm³ Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000041 CAS Number: 75-16-1 EINECS: 200-844-1 UN: 1928 ADR: 4.3,I |
Specification Test Results
Appearance | Clear to hazy gray to brown to black Solution (May develop some turbidity or precipitate) |
Titration Grignard | 2.85 to 3.20 M |
Melting point | 120 to 124 °C |
Solubility | (5 % in ethanol) Clear solution |
Residue after ignition | ≤0.1 % |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...