Sodium triacetoxyborohydride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
29182-1000
29182-5000
29182-0250
TS29182-1000EA
207
USD
TS29182-1000
TS29182-5000
TS29182-0250
Sodium triacetoxyborohydride 97%
Sodium triacetoxyborohydride
Formula:
C₆H₁₀BNaO₆ Melting Pt: 114…118 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012211 CAS Number: 56553-60-7 UN: 3396 ADR: 4.3,II |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Assay | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...