Resazurin sodium salt, pure, high purity biological stain
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin
18990-0050
18990-0010
18990-0250
TS18990-0050EA
84
USD
TS18990-0050
TS18990-0010
TS18990-0250
Resazurin sodium salt, pure, high purity biological stain
Resazurin sodium salt
Formula:
C₁₂H₆NNaO₄ MW: 251.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005036 CAS Number: 62758-13-8 EINECS: 263-718-5 |
Specification Test Results
Appearance | Dark green to black Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥75 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...