Boron trifluoride - methanol complex 12% 1.5 M in methanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

44527-8000 44527-1000
TS44527-8000EA 224 USD
TS44527-8000 TS44527-1000
Boron trifluoride - methanol complex 12% 1.5 M in methanol, AcroSeal™
Boron trifluoride - methanol complex
Formula: CH₄BF₃O
MW: 99.85
Boiling Pt: 65 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.87 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00071635
CAS Number: 373-57-9
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Assay 10 to 15 % w/v (BF3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR