Boron trifluoride - methanol complex 12% 1.5 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

40276-0010 40276-5000 402761000
TS40276-0010EA 176 USD
TS40276-0010 TS40276-5000 TS402761000
Boron trifluoride - methanol complex 12% 1.5 M in methanol
Boron trifluoride - methanol complex
Formula: CH₄BF₃O
MW: 99.85
Melting Pt: –98 °C
Density: 0.87 g/cm³
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to amber Solution
Titration with NaOH 10 to 15 % w/v
Refractive index 1.5250 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR