1,2,4-Triazole 99.5%

Supplier: Thermo Scientific Chemicals
Warning

13928-5000 13928-0050 13928-0250 13928-1000
TS13928-5000EA 417 USD
TS13928-5000 TS13928-0050 TS13928-0250 TS13928-1000
1,2,4-Triazole 99.5%
1,2,4-Triazole
Formula: C₂H₃N₃
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: 119…122 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00005228
CAS Number: 288-88-0
EINECS: 206-022-9
Merck Index: 12,09735

Order Now

Specification Test Results

Appearance White Powder and/or flakes
Infrared spectrum Conforms
Melting point 119 to 122 °C
Water ≤0.2 % (K.F.)
HPLC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR