Fuchsin (acid), pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CI 42685

22790-1000 22790-0250 22790-5000
TS22790-1000EA 229 USD
TS22790-1000 TS22790-0250 TS22790-5000
Fuchsin (acid), pure, high purity biological stain
Fuchsin (acid)
Formula: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109

Order Now

Specification Test Results

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
UV Lambda max 544 to 550 nm (in pH 3 acidified water)
Absorption Ratio P-15/P+15 1 to 1.4
Specific absorbance A (1%/1cm) ≥700 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR