4-Bromostyrene 96% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-4-vinylbenzene
15428-1000
15428-0100
15428-0250
TS15428-1000EA
689
USD
TS15428-1000
TS15428-0100
TS15428-0250
4-Bromostyrene 96% stabilized
4-Bromostyrene
Formula:
C₈H₇Br MW: 183.05 g/mol Boiling Pt: 88…89 °C (16 mmHg) Melting Pt: 4.5 °C Density: 1.4 g/cm³ Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000110 CAS Number: 2039-82-9 EINECS: 218-022-6 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5920 to 1.5960 (20°C, 589 nm) |
GC | ≥95 % |
Stabilizer | 0.05 to 0.1 % 4-tert-Butylcatechol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...