Ethylene di(acetate) 99%

Supplier: Thermo Scientific Chemicals

17622-0100 17622-0025 17622-5000
TS17622-0100EA 349 USD
TS17622-0100 TS17622-0025 TS17622-5000
Ethylene di(acetate) 99%
Ethylene di(acetate)
Formula: C₆H₁₀O₄
Boiling Pt: 186…187 °C (760 mmHg)
Melting Pt: –41 °C
Density: 1.12 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00008718
CAS Number: 111-55-7
EINECS: 203-881-1
Merck Index: 13,03833

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4142 to 1.4162 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR